Learn More
Fisherbrand™ pH Test Paper Rolls
General purpose pH papers in efficient 15 ft. rolls
Supplier: Fisherbrand™ B01SHTRG125140SRD
Description
General purpose pH papers in efficient 15 ft. rolls
- Convenient dispenser allows you to select the right amount of pH paper for each application
- Easy-to-use—Dip paper in solution to be tested and match to color chart immediately
- Highly accurate and well suited for most applications
- Each SKU is packed in box of five clear plastic dispensers
- Cat. No. 13-640-507 is a pH paper testing set that includes shrink wrapped pack of seven dispensers (one of each SKU)
Order Info
15 ft. Roll provides about 100 tests.
Chemical Identifiers
| Convenient dispenser allows to select the right amount of pH paper for each application, highly accurate and well suited for most applications | |
| Room Temperature | |
| pH Test Paper |
Specifications
| 12.5 to 14 | |
| 0.5 | |
| Room Temperature |
| Convenient dispenser allows to select the right amount of pH paper for each application, highly accurate and well suited for most applications | |
| 100 Tests/Roll, 500 Total | |
| pH Test Paper |