Learn More
Fisherbrand™ Permanent Markers
Write on all surfaces
 View more versions of this product
Catalog No. 13383D
Only null left
Description
Description
Fisherbrand™ Permanent Marking Pens write on glass, film, wood, plastic, metal, rubber, and porcelain. Perfect for marking sample bags, microscope or photographic slides, laboratory glassware and more. Write on all surfaces.
  • Produce precise, ultrathin lines 0.3mm wide
  • Waterproof and fadeproof, permanent ink dries almost instantly
  • An airtight ring seal cap prevents dry-out and ensures instant writing even after lengthy storage
  • Unique stainless-steel long leg allows precise writing in a confined space
Order Info
Assorted includes six opaque black, two vivid red, two blue, and two green pens
Specifications
Chemical Identifiers
Permanent Pen
Stainless Steel
0.3 mm
0.3 mm Ultra Fine
Perfect marking pens for lab samples, microscope slides, photographic slides, glass and plastic beakers, and plastic sample bags
Specifications
Permanent Pen
Red
Perfect marking pens for lab samples, microscope slides, photographic slides, glass and plastic beakers, and plastic sample bags
0.3 mm
0.3 mm Ultra Fine
Stainless Steel
6 Pk.
Product Suggestions
Product Suggestions