Learn More
Fisherbrand™ Nonsterile Threaded End Microcentrifuge Tubes without Caps
Choose skirted- or conical-bottom style.
$90.00 - $124.70
| Catalog Number | Color | Capacity (Metric) | Max. RCF | Bottom Shape | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Color | Capacity (Metric) | Max. RCF | Bottom Shape | Price | Quantity & Availability | |||
02681333
![]() |
Natural | 0.5 mL | 30,000 x g | Skirted |
Pack of 500 Each for $90.00
Case of 10 Pack for $900.00
|
|
|||
02681334
![]() |
Natural | 0.5 mL | 25,000 x g | Conical |
Pack of 500 Each for $101.85
Case of 10 Pack for $1,018.50
|
|
|||
02681337
![]() |
Amber | 0.5 mL | 30,000 x g | Skirted |
Pack of 500 Each for $98.52
Case of 10 Pack for $985.20
|
|
|||
02681335
![]() |
Amber | 0.5 mL | 25,000 x g | Conical |
Pack of 500 Each for $98.41
Case of 10 Pack for $984.10
|
|
|||
02681338
![]() |
Natural | 1.5 mL | 18,000 x g | Skirted |
Pack of 500 Each for $122.70
Case of 10 Pack for $1,227.00
|
|
|||
02681339
![]() |
Natural | 1.5 mL | 30,000 x g | Conical |
Pack of 500 Each for $91.85
Case of 10 Pack for $918.50
|
|
|||
02681340
![]() |
Amber | 1.5 mL | 30,000 x g | Conical |
Pack of 500 Each for $95.59
Case of 10 Pack for $955.90
|
|
|||
02681342
![]() |
Amber | 1.5 mL | 18,000 x g | Skirted |
Pack of 500 Each for $124.70
Case of 10 Pack for $1,247.00
|
|
|||
02681343
![]() |
Natural | 2 mL | 18,000 x g | Skirted |
Pack of 500 Each for $95.52
Case of 10 Pack for $955.20
|
|
|||
02681344
![]() |
Natural | 2 mL | 25,000 x g | Conical |
Pack of 500 Each for $91.85
Case of 10 Pack for $918.50
|
|
|||
02681345
![]() |
Amber | 2 mL | 25,000 x g | Conical |
Pack of 500 Each for $95.74
Case of 10 Pack for $957.40
|
|
|||
02681346
![]() |
Amber | 2 mL | 18,000 x g | Skirted |
Pack of 500 Each for $101.56
Case of 10 Pack for $1,015.60
|
|
|||
Description
Choose skirted- or conical-bottom style.
- Threaded to accept colored screw caps (sold separately)
- Siliconized/low-retention conical-bottom tubes also available
Specifications
| Non-sterile | |
| Screw Cap |