missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Microcentrifuge Tubes with Tether Cap

Reduce sample contamination by keeping cap with tube
$456.89 - $925.56
Specifications
| Closure Material | Polypropylene |
|---|---|
| Autoclavable | Autoclavable |
| Color | Natural |
| Material | Polypropylene |
| For Use With (Application) | Suitable for vapor-phase liquid nitrogen storage, boiling and autoclaving |
| Catalog Number | Sterility | Capacity (Metric) | Bottom Shape | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Sterility | Capacity (Metric) | Bottom Shape | Price | Quantity & Availability | ||||
|
02707350
|
Non-sterile | 0.5 mL | Round |
Pack of 500 Each for $607.52
Case of 10 Pack for $6,075.20
|
|
||||
|
02707354
|
Non-sterile | 2 mL | Round |
Pack of 500 Each for $456.89
Case of 10 Pack for $4,568.90
|
|
||||
02707352
![]() |
Non-sterile | 1.5 mL | Round |
Pack of 500 Each for $467.41
Case of 10 Pack for $4,674.10
|
|
||||
02707351
![]() |
Sterile | 0.5 mL | Round |
Pack of 500 Each for $914.89
Case of 5 Pack for $4,574.45
|
|
||||
02707353
![]() |
Sterile | 1.5 mL | Round |
Pack of 500 Each for $807.44
Case of 5 Pack for $4,037.20
|
|
||||
02707355
![]() |
Sterile | 2 mL | Round |
Pack of 500 Each for $530.74
Case of 5 Pack for $2,653.70
|
|
||||
02707356
![]() |
Non-sterile | 0.5 mL | Skirted |
Pack of 500 Each for $478.56
Case of 10 Pack for $4,785.60
|
|
||||
|
02707357
|
Sterile | 0.5 mL | Skirted |
Pack of 500 Each for $852.48
Case of 5 Pack for $4,262.40
|
|
||||
|
02707358
|
Non-sterile | 1.5 mL | Skirted |
Pack of 500 Each for $551.70
Case of 10 Pack for $5,517.00
|
|
||||
|
02707359
|
Sterile | 1.5 mL | Skirted |
Pack of 500 Each for $925.56
Case of 5 Pack for $4,627.80
|
|
||||
02707360
![]() |
Non-sterile | 2 mL | Skirted |
Pack of 500 Each for $472.11
Case of 10 Pack for $4,721.10
|
|
||||
02707361
![]() |
Sterile | 2 mL | Skirted |
Pack of 500 Each for $816.41
Case of 5 Pack for $4,082.05
|
|
||||
Description
Reduce sample contamination by keeping cap with tube
Natural-colored polypropylene tubes include leash-and-loop tether. Ultragrade polypropylene caps are fitted with ethylene/propylene O-rings. Available sterile or nonsterile with round or skirted bottoms.
- Removable tether rotates freely for easy opening and closing
- O-ring forms positive seal with tube to reduce leakage
- Withstand 20,000xG
- Suitable for vapor-phase liquid nitrogen storage, boiling and autoclaving
- RNase and DNase free
Order Info
Nonsterile tubes come separate from tether caps; sterile tubes come attached; 500 per pack
Specifications
| Polypropylene | |
| Natural | |
| Suitable for vapor-phase liquid nitrogen storage, boiling and autoclaving | |
| Tether Cap |
| Autoclavable | |
| Polypropylene | |
| Bag | |
| 20,000 x g |