missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Locking-Lid Microcentrifuge Tubes with Polypropylene Snap-Cap™
GREENER_CHOICE

Catalog No. 02681300
Change view
Click to view available options
Color:
Natural
Capacity (Metric):
0.6 mL
1.5 mL
Max. RCF:
26,000 x g
30,000 x g
Packaging:
Bag
Single Wrap
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Capacity (Metric) Max. RCF Packaging
02681300 Natural 0.6 mL 30,000 x g Single Wrap
02681273 Natural 0.6 mL 30,000 x g Bag
02681284 Natural 1.5 mL 26,000 x g Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02681300
Only null left

Convenient, pierceable locking lids snap into place with one-handed operation

  • Polypropylene tubes with flat tops
  • Ideal for sample-handling applications, including centrifugation, boiling and freezing of specimens, incubation and storage
  • With graduation marks for volume estimation
  • RNase and DNase free

Centrifuges that develop excessive heat may damage this tube.

Specifications

Certifications/Compliance RNase, DNase-free
Closure Material Polypropylene
Autoclavable Autoclavable
Capacity (Metric) 0.6 mL
Color Natural
Material Polypropylene
For Use With (Application) Ideal for sample-handling applications, including centrifugation, boiling and freezing of specimens, incubation and storage
Packaging Single Wrap
Sterility Sterile
Closure Type Snap Cap
Max. RCF 30,000 x g
Graduations 0.1 mL
Show More Show Less