Learn More
Fisherbrand™ Large Tip Opening Serological Pipets
Ideal for viscous solutions and tissue culture work.
 View more versions of this product
Catalog No. 13-671-108C
Only null left
Explore available promotions
Description
Description
Ideal for viscous solutions and tissue culture work.
  • Tops formed for cotton plugging (plugs not included) will fit cotton plugging machines 1mL size has approximately 0.1 in. (2.5mm) tip opening; other sizes have approximately 0.12 in. (3mm) tip opening
  • Accuracy printed on each pipet
  • Borosilicate glass with permanent amber markings
  • Tuf-Temp™ treated to reduce chipping
Compliance
Calibrated To Deliver/Blow Out.
Specifications
Specifications
±0.1 mL
Borosilicate Glass
0.1 mL
5 mL
Large
Yes
Blue
Serological Pipet
Product Suggestions
Product Suggestions