missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Large Biohazard Labels

Bright red fluorescent labels display the universal biohazard symbol for easy identification.
$10.72 - $13.99
Specifications
| Certifications/Compliance | OSHA |
|---|---|
| Color | Black, Red |
| Format | Label |
| Material | Paper |
| For Use With (Application) | Biohazard |
| Catalog Number | Mfr. No. | Quantity | Size (English) | Size (Metric) | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Size (English) | Size (Metric) | Price | Quantity & Availability | |||
18999935
![]() |
Fisherbrand™
LBH 23 |
640/Pk. | 0.5 x 1.5 in. | 1.3 x 3.8 cm |
Each for $10.72
|
|
|||
18999936
![]() |
Fisherbrand™
LBH 24 |
320/Pk. | 3 x 3 in. | 7.6 x 7.6 cm |
Each for $13.99
|
|
|||
Description
Bright red fluorescent labels display the universal biohazard symbol for easy identification.
Strong adhesive ensures that labels remain in place.
Compliance
Meet OSHA requirements.
Specifications
| OSHA | |
| Label | |
| Biohazard |
| Black, Red | |
| Paper |