missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ InkJet Plus Microscope Slides

Catalog No. p-4876638
Change view
Click to view available options
Color:
Aqua
Blue
Green
Lilac
Pink
Tan
White
Yellow
Corner Style:
45°
90°
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Corner Style
12550112 Pink 90°
12550111 Green 90°
12550109 White 90°
12550110 Blue 90°
12550113 Yellow 90°
12550114 Aqua 90°
12550116 Lilac 90°
12550117 Tan 90°
22042925 Blue 45°
22042924 White 45°
22042927 Pink 45°
22042929 Aqua 45°
22042931 Lilac 45°
22042932 Tan 45°
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 12550112
Only null left

Superfrost™/ColorFrost™ microscope slides designed for use with Leica™ Microsystems IP S inkjet printer

  • Printing instruments use chemical-resistant ink and print directly on Superfrost/ColorFrost tab of the slide
  • For histology, cytology, hematology, and microbiology applications
  • Clipped 45° or straight 90° corners
  • Include a plus (+) charge symbol
  • Frosted tabs in nine different colors

Specifications

Frosted Tabs
Color Pink
Corner Style 90°