missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Infecon™ 1000 Infectious Substance Shipper Absorbent Sheets
For use with Fisherbrand Infecon Infectious Substance Shippers
Specifications
| Quantity | 100 Sheets |
|---|---|
| Includes | All components needed for shipping infectious substances; with step-by-step instructions to aid compliance with D.O.T. regulations |
| Applications | For infectious substance shipper/Dry ice |
| Type | Absorbent Sheet |
Description
Certifications
UN Certified
Compliance
Complies with ICAO/IATA Packing Instruction 602.
Specifications
| 100 Sheets | |
| All components needed for shipping infectious substances; with step-by-step instructions to aid compliance with D.O.T. regulations | |
| For infectious substance shipper/Dry ice | |
| Absorbent Sheet |