missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Infecon™ 1000 Infectious Substance Shipper Absorbent Sheets

Catalog No. 19075383C
Change view
Click to view available options
Type:
Absorbent Sheet
Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Size
19075383C Absorbent Sheet 250mL
19075383B Absorbent Sheet 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19075383C Supplier Fisherbrand™ Supplier No. FSB1004

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with Fisherbrand Infecon Infectious Substance Shippers

Certifications

UN Certified

Compliance

Complies with ICAO/IATA Packing Instruction 602.

Specifications

Quantity 100 Sheets
Includes All components needed for shipping infectious substances; with step-by-step instructions to aid compliance with D.O.T. regulations
Applications For infectious substance shipper/Dry ice
Size 250mL
Type Absorbent Sheet