missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ GHS Labeled Right-to-Know Safety-Vented Wash Bottles

Catalog No. p-4953403
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
Distilled Water
Isopropanol
Methanol
Closure Color:
Blue
Green
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Chemical Label Closure Color
11865153 500 mL Distilled Water Blue
11865167 1000 mL Isopropanol Yellow
11865168 1000 mL Methanol Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11865153 Supplier Fisherbrand™ Supplier No. 124163104
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Unique valve prevents pressure build-up, eliminating potentially dangerous chemical drips and allows dispensing of wash bottle in upright or inverted position.

  • Pre-printed bottles with chemical name and formula, GHS pictogram and signal word; National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
  • Translucent LDPE bottles, Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 nm
  • Wide mouth 53 mm (2 in.) for easy refilling
  • Color-coded polypropylene closures aid in quick and easy identification
  • Assortment bag includes one each: Acetone, Isopropanol, Bleach and Ethanol
  • Bottle with Label: TOLUENE, Red LDPE bottle, does not include vented valve
  • Bottle with Label: DEIONIZED H2O, DISTILLED H20 and WATER, does not indicate hazardous chemical and does not have or require GHS pictogram or signal word
  • Bottle with Label: METHYL ETHYL KETONE, does not include vented valve

Compliance

Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA’s HazCom 2012 requirements – as pertains to Workplace Labels.

Specifications

Capacity 500 mL
Capacity (Metric) 500 mL
Color Natural
Material LDPE
Quantity 3/Pk.
Sterility Non-sterile
Closure Color Blue
Closure Included Yes
Graduated No
Mouth Wide
Chemical Label Distilled Water
Closure Material Polypropylene
Closure Size 53 mm
Closure Type Cap
Through the Closure Dispensing Yes
Autoclavable Not Autoclavable
Capacity (English) 16.91 oz.
Shape Cylindrical
Disposable Reusable
Product Type Safety-vented Wash Bottle
Show More Show Less