missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PurSwab Foam Swabs

Flexible foam applicators. Tip is comprised of an open-cell polyurethane foam. Ideal for a variety of cleaning applications.

$30.86 - $56.34

Specifications

Certifications/Compliance ISO 9001:2008, ISO 13485:2003, FDA GMP's
Product Type Swab
Grade Medical
Sterility Non-sterile
View More Specs

Products 4
Catalog Number Length (Metric) Handle Material Price Quantity & Availability  
Catalog Number Length (Metric) Handle Material Price Quantity & Availability  
149603E
View Documents Promotion Details
7.14 cm Polypropylene
Pack of 50 Each for $30.86
Case of 10 Pack for $308.60
Only null left
 
149603G
View Documents Promotion Details
14.9 cm Wood
Pack of 50 Each for $44.34
Case of 10 Pack for $443.40
Only null left
 
149603H
View Documents Promotion Details
15.2 cm Wood
Pack of 50 Each for $45.00
Case of 10 Pack for $450.00
Only null left
 
149603L
View Documents Promotion Details
13.1 cm Polypropylene
Pack of 50 Each for $56.34
Case of 10 Pack for $563.40
Only null left
 
Description

Description

Flexible mini-tip foam applicators. Tip is comprised of an open-cell polyurethane foam. Ideal for a variety of cleaning applications.

  • With polypropylene or wood handles
  • Nonlinting and nonscratching

Applications:

May be used with alcohol, acetone, oils, greases, and solvents

Specifications

Specifications

ISO 9001:2008, ISO 13485:2003, FDA GMP's
Swab
Medical
Non-sterile
Videos
SDS
Documents

Documents