missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Flat and Domed 8-Cap Strips, Flat Cap Strips

Catalog No. 14230231
Change view
Click to view available options
Format:
Domed Cap Strips
Flat Cap Strips
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
14230231 Domed Cap Strips
14230230 Flat Cap Strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14230231 Supplier Fisherbrand™ Supplier No. 14230231

Suitable for use with 96-well PCR plates and PCR tube strips

  • Eight caps per strip
  • Cleanroom produced
  • Virgin polypropylene
  • Flat caps offer large, labeled end tabs for easy removal and strip orientation

Specifications

Material Polypropylene
Color Natural
Format Domed Cap Strips
For Use With (Equipment) 96-well PCR Plates or PCR Tube Strips