missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Flat and Domed 8-Cap Strips, Flat Cap Strips
Description
Suitable for use with 96-well PCR plates and PCR tube strips
- Eight caps per strip
- Cleanroom produced
- Virgin polypropylene
- Flat caps offer large, labeled end tabs for easy removal and strip orientation
Specifications
Specifications
| Material | Polypropylene |
| Color | Natural |
| Format | Flat Cap Strips |
| For Use With (Equipment) | 96-well PCR Plates or PCR Tube Strips |