Learn More
Fisherbrand™ Dissecting Blunt-Pointed Forceps
Fisherbrand Blunt-Pointed Dissection Forceps are constructed from heavy duty stainless-steel, perfect for long lasting use.
$4.38 - $5.66
Specifications
Product Type Dissecting Forcep
Autoclavable Autoclavable
Certifications/Compliance ASTM
Includes Guide pins
Latex Free Yes
View More Specs
Products 3
Catalog Number Length (Metric) Tip Style Material Price Quantity & Availability  
Catalog Number Length (Metric) Tip Style Material Price Quantity & Availability  
08895
View Documents Promotion Details
10.46 cm Straight, serrated, blunt point Chrome-plated Steel
Each for $4.90
Case of 10 Each for $49.00
Only null left
08890
View Documents Promotion Details
12.7 cm Straight, serrated, blunt point Chrome-plated Steel
Each for $4.38
Case of 10 Each for $43.80
Only null left
08887
View Documents Promotion Details
12.7 cm Curved, serrated, blunt point Nickel-plated Steel
Each for $5.66
Case of 12 Each for $67.92
Only null left
Description
Description
Fisherbrand Blunt-Pointed Dissection Forceps are constructed from heavy duty stainless-steel, perfect for long lasting use.
  • Fisherbrand™ Medium-Pointed Forceps are designed to exceed perfromance expectations and manufactured to exact standards
  • Tips grasp tissues and materials
  • Tips meet precisely when closed
  • Patterned-grip handles with guide pins (08-882 and 08-885)
  • Smooth handle without guide pins (08-900)
  • Passivated for corrosion resistance
  • ASTM™ standard stainless steel construction
  • Latex-free
  • Can be sterilized by Eto/gamma radiation
  • Rockwell hardness (HRC): 40-45
  • Fully autoclavable
Specifications
Specifications
Dissecting Forcep
ASTM
Yes
Autoclavable
Guide pins