missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Disposable Divided Well Pipette Basins

Catalog No. p-4893799
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Packaging:
10/Bag
Individually Wrapped
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Packaging
13681505 Sterile Individually Wrapped
13681506 Sterile 10/Bag
13681507 Non-sterile 10/Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13681505 Supplier Fisherbrand™ Supplier No. 13681505
Only null left

Disposable basins offer convenience, fast cleanup

  • Easy-pour corners for quick disposal of excess sample solution
  • Gamma-irradiated sterilized basins
  • Packaged in easy-to-open bags to reduce contamination risks

  • Divided well design allows for the pipetting of two reagents at the same time
  • Dimensions: 5.7L × 2.4W × 0.8 in.H (14.5 × 6.3 × 2.1cm)

Specifications

Description Gamma-irradiated sterilized
Capacity (Metric) 25 mL
Dimensions (L x W x H) 5.7 x 2.4 x 0.8 in. (14.5 x 6.3 x 2.1cm)
Material Polystyrene
Sterility Sterile
Type Pipetting Reservoir
Height (English) 0.8 in.
Height (Metric) 2.1 cm
Length (English) 5.7 in.
Length (Metric) 14.5 cm
Packaging Individually Wrapped
Width (English) 2.4 in.
Width (Metric) 6.3 cm
Disposable Single-use
Show More Show Less