Learn More
Fisherbrand™ Freeze Cube XL
Freeze Cube XL is a lightweight, ice free and easy to carry cooling box that offers low temperature protection for biological samples all day long while working on the bench.
$976.07 - $8525.66
Specifications
Dimensions (L x W x H) 152 x 170 x 123 mm
View More Specs
Products 6
Catalog Number Mfr. No. Color Includes Price Quantity  
Catalog Number Mfr. No. Color Includes Price Quantity  
13131004
View Documents
N/A Purple 30-well Cooling Blocks
Each for $1,019.31
Case of 8 Each for $8,154.48
Only null left
13131005
View Documents
N/A Green 30-well Cooling Blocks
Each for $1,019.31
Case of 8 Each for $8,154.48
Only null left
13131006
View Documents
N/A Blue 30-well Cooling Blocks
Each for $1,019.31
Case of 8 Each for $8,154.48
Only null left
13131007
View Documents
N/A Purple 96-well Cooling Blocks
Case of 8 Each for $8,525.66
Only null left
13131008
View Documents
N/A Green 96-well Cooling Blocks
Each for $976.07
Case of 8 Each for $7,808.56
Only null left
13131009
View Documents
N/A Blue 96-well Cooling Blocks
Each for $1,065.66
Case of 8 Each for $8,525.28
Only null left
Description
Description
This cooling box is ideal for low temperature protection and collection of biological samples, can also be used to preserve unstable nucleic acid samples while working on the bench.
Specifications
Specifications
152 x 170 x 123 mm
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'