Learn More
Fisherbrand™ Freeze Cube XL
Freeze Cube XL is a lightweight, ice free and easy to carry cooling box that offers low temperature protection for biological samples all day long while working on the bench.
 View more versions of this product
Catalog No. 13131009
Only null left
Explore available promotions
Description
Description
This cooling box is ideal for low temperature protection and collection of biological samples, can also be used to preserve unstable nucleic acid samples while working on the bench.
Specifications
Specifications
Blue
96-well Cooling Blocks
152 x 170 x 123 mm
Product Suggestions
Product Suggestions