Learn More
Fisherbrand™ Cleanroom Notebooks, Latex-Free
Used for documentation in critical environments requiring low particulation and ESD generation. Fisherbrand™ Cleanroom Notebooks come in a variety of sizes.
Supplier:  Fisherbrand™ 01NB3X5LF
 Afficher plus de versions de ce produit
Numéro de catalogue. 18001546
Il en reste null
Description
Description
  • For documentation in critical environments
  • Low particulate and ESD generation
  • Printed on #22 paper with college rule lines.
  • Semi-transparent high density polyethylene (HDPE) covers
  • 100 pages/50 sheets
  • Spiral bound
Compliance
Class 10 Certification
Spécifications
Identifiants chimiques
Latex-free
Spécifications
Latex-free
3.5 x 5 in.
22#
College Ruled
Notebook
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS