Learn More
Fisherbrand™ Cleanroom Notebooks, Latex-Free
Used for documentation in critical environments requiring low particulation and ESD generation. Fisherbrand™ Cleanroom Notebooks come in a variety of sizes.
Supplier: Fisherbrand™ 01NB5.5X8.5GRIDLF
Description
- For documentation in critical environments
- Low particulate and ESD generation
- Printed on #22 paper with college rule lines.
- Semi-transparent high density polyethylene (HDPE) covers
- 100 pages/50 sheets
- Spiral bound
Compliance
Class 10 Certification
Chemical Identifiers
| Latex-free |
Specifications
| Latex-free | |
| 5.5 x 8.5 in. | |
| 22# |
| Grid | |
| Notebook |