Learn More
Fisherbrand™ Cleanroom Notebooks, Latex-Free
Catalog No. 18001503
Change view
Click to view available options
Dimensions (L x W):
5.5 x 8.5 in.
5.875 x 8.25 in.
8.25 x 11.75 in.
8.3 x 11.7 in.
3.5 x 5 in.
8.5 x 11 in.
Ruled:
College Ruled
Grid
Yes
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Ruled
18001503 5.5 x 8.5 in. College Ruled
18003059 8.3 x 11.7 in. Yes
18001542 8.5 x 11 in. College Ruled
18001543 8.5 x 11 in. Grid
18001545 5.5 x 8.5 in. Grid
18001546 3.5 x 5 in. College Ruled
18003065 8.25 x 11.75 in. College Ruled
18003066 5.875 x 8.25 in. College Ruled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18001503 Supplier Fisherbrand™ Supplier No. 18001503
Used for documentation in critical environments requiring low particulation and ESD generation. Fisherbrand™ Cleanroom Notebooks come in a variety of sizes.
  • For documentation in critical environments
  • Low particulate and ESD generation
  • Printed on #22 paper with college rule lines.
  • Semi-transparent high density polyethylene (HDPE) covers
  • 100 pages/50 sheets
  • Spiral bound
Specifications
Material Latex-free
Ruled College Ruled
Cover Material HDPE
Description 30/case
Dimensions (L x W) 5.5 x 8.5 in.
Certifications/Compliance ISO Clean 10