missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Chemical Yellow Absorbent Pad

Catalog No. 19140918
Change view
Click to view available options
Capacity (English):
22 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19140918 22 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19140918 Supplier Fisherbrand™ Supplier No. FSCCH1519M100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Color-coded for easy identification of different types of spills and hazards

  • Surfactant-treated, brightly colored absorbent picks up a wide range of aggressive chemicals while remaining chemically inert
  • Heavy weight - specifically designed for continuous leaks and larger spills so you use less material
  • BRIGHTSORB™ High Visibility Series feature safety signage and absorbency in one
  • Perforations provide cost control and reduce waste by allowing you to use only what you need

Compliance

OSHA 29 CFR 1910.22(a)(2); EPA 40 CFR 112.7(c)(1)(vii); OSHA 29 CFR 1910,120(j)(1)(vii).

Specifications

Color Yellow
Capacity (English) 22 gal.
Capacity (Metric) 83.2 L
Material Polypropylene
Type Chemical Absorbent Pad
Packaging Quantity 100/Cs.
Weight Capacity Heavyweight
Dimensions (L x W) 48.2 x 38.1 cm (19 x 15 in.)
Length (English) 19 in.
Length (Metric) 48.2 cm
Width (English) 15 in.
Width (Metric) 38.1 cm
Show More Show Less