missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cell Spreaders

Simplify bacterial culture work and prevent agar surface tears.

$144.04 - $378.74

Specifications

Autoclavable Autoclavable
Material Polypropylene
Product Type Cell Spreader
For Use With (Application) Simplify bacterial culture work and prevent agar surface tears, easy to manipulate around curves and into corners of prepared plates
View More Specs

Products 2
Catalog Number Color Sterility For Use With (Equipment) Price Quantity & Availability  
Catalog Number Color Sterility For Use With (Equipment) Price Quantity & Availability  
0554110
View Documents Promotion Details
Blue Non-sterile Prepared Biplates
Pack of 25 Each for $144.04
Case of 4 Pack for $576.16
Only null left
 
087735
View Documents Promotion Details
Yellow Sterile 100 to 150 mm Prepared Plates
Pack of 25 Each for $378.74
Case of 4 Pack for $1,514.96
Only null left
 
Description

Description

  • Autoclavable, nonbreakable and reusable polypropylene
  • Use 30mm wide spreaders for prepared biplates
  • Use 60mm wide spreaders for 100 to 150mm prepared plates

Order Info

Sold in packs of 25; sterile spreaders are individually wrapped.

Specifications

Specifications

Autoclavable
Polypropylene
Cell Spreader
Simplify bacterial culture work and prevent agar surface tears, easy to manipulate around curves and into corners of prepared plates
Videos
SDS
Documents

Documents