missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cell Spreaders

Catalog No. 087735
Change view
Click to view available options
Color:
Blue
Yellow
Sterility:
Non-sterile
Sterile
For Use With (Equipment):
100 to 150 mm Prepared Plates
Prepared Biplates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility For Use With (Equipment)
087735 Yellow Sterile 100 to 150 mm Prepared Plates
0554110 Blue Non-sterile Prepared Biplates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 087735

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Simplify bacterial culture work and prevent agar surface tears.

  • Autoclavable, nonbreakable and reusable polypropylene
  • Use 30mm wide spreaders for prepared biplates
  • Use 60mm wide spreaders for 100 to 150mm prepared plates

Order Info

Sold in packs of 25; sterile spreaders are individually wrapped.

Specifications

Autoclavable Autoclavable
For Use With (Equipment) 100 to 150 mm Prepared Plates
Sterility Sterile
Material Polypropylene
Product Type Cell Spreader
Color Yellow
For Use With (Application) Simplify bacterial culture work and prevent agar surface tears, easy to manipulate around curves and into corners of prepared plates