Learn More
Fisherbrand™ Brix Hydrometers
Used to measure the concentration of sugar solutions in refineries.
Description
- Calibrated at 20°C
- Permanently marked with individual serial numbers
For measuring the concentration of sugar solutions in refineries
Order Info
A certificate of calibration is available; simply add the letter C to the end of the catalog number when ordering.
Certifications
NIST™ traceable. A certificate is provided to indicate traceability to standards provided by NIST™.
Compliance
Accuracy meets or exceeds ASTM™ E 100 specifications, calibrated according to ASTM™ E 126 specifications.
Specifications
| 0.1 | |
| 300 mm | |
| 0 to 12° | |
| Measuring the concentration of Sugar solutions in refineries |
| NIST, ASTM E100, ASTM E126 | |
| 300mL BSG cylinder (08-530J) | |
| Soda Lime Glass | |
| 250 mL |