Learn More
Fisherbrand™ Brix Hydrometers
Used to measure the concentration of sugar solutions in refineries.
$43.35 - $54.47
Specifications
| For Use With (Application) | Measuring the concentration of Sugar solutions in refineries |
|---|---|
| Volume (Metric) | 250 mL |
| Catalog Number | Length (Metric) | For Use With (Equipment) | Brix Range | Increments | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Length (Metric) | For Use With (Equipment) | Brix Range | Increments | Price | Quantity & Availability | |||
|
11608C
|
300 mm | 300mL BSG cylinder (08-530J) | 19 to 31° | 0.1 |
Each for $54.40
|
|
|||
|
11610B
|
375 mm | 1 L Heavy wall cylinder (08-532D) | 0 to 50° | 0.5 |
Each for $54.47
|
|
|||
|
11610A
|
300 mm | 300mL BSG cylinder (08-530J) | 35 to 70° | 0.5 |
Each for $49.13
|
|
|||
|
11608E
|
300 mm | 300mL BSG cylinder (08-530J) | 39 to 51° | 0.1 |
Each for $54.40
|
|
|||
|
11608D
|
300 mm | 300mL BSG cylinder (08-530J) | 29 to 41° | 0.1° |
Each for $54.40
|
|
|||
|
11608G
|
300 mm | 300mL BSG cylinder (08-530J) | 59 to 71° | 0.1° |
Each for $54.47
|
|
|||
|
11608A
|
300 mm | 300mL BSG cylinder (08-530J) | 0 to 12° | 0.1 |
Each for $50.84
|
|
|||
|
11608B
|
300 mm | 300mL BSG cylinder (08-530J) | 9 to 21° | 0.1 |
Each for $50.84
|
|
|||
|
11610
|
295 mm | 300mL BSG cylinder (08-530J) | 0 to 35° | 0.5° |
Each for $43.35
|
|
|||
Description
- Calibrated at 20°C
- Permanently marked with individual serial numbers
For measuring the concentration of sugar solutions in refineries
Certifications
NIST™ traceable. A certificate is provided to indicate traceability to standards provided by NIST™.
Compliance
Accuracy meets or exceeds ASTM™ E 100 specifications, calibrated according to ASTM™ E 126 specifications.
Specifications
| Measuring the concentration of Sugar solutions in refineries | |
| 250 mL |