missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Bottle-Top Dispenser Anti-tip Reservoirs

Side neck allows filling of reservoir without removing the dispenser

$306.96 - $729.00

Specifications

Product Type Anti-Tip Reservoir
Autoclavable Not Autoclavable
Color Amber
Material Borosilicate Glass, Nylon Side Filter Neck
For Use With (Equipment) Bottletop Dispenser
View More Specs

Products 2
Catalog Number Mfr. No. Volume (Metric) Diameter (Metric) Neck Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Diameter (Metric) Neck Price Quantity & Availability  
13681519
View Documents Promotion Details
Fisherbrand™
2.5 L AMBER RESERVOIR
2500 mL 45 mm
Each for $729.00
Only null left
 
13681518
View Documents Promotion Details
Fisherbrand™
1.4 L AMBER RESERVOIR
1400 mL 30 mm
Each for $306.96
Only null left
 
Description

Description

 

  • Amber borosilicate glass
  • Nylon side filter neck
  • Low profile and wide footprint for extra stability

 

Specifications

Specifications

Anti-Tip Reservoir
Amber
Bottletop Dispenser
Not Autoclavable
Borosilicate Glass, Nylon Side Filter Neck
Side filler neck, low profile and wide footprint
Videos
SDS
Documents

Documents