missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Bottle-Top Dispenser Anti-tip Reservoirs

Catalog No. 13681519
Change view
Click to view available options
Volume (Metric):
1400 mL
2500 mL
Diameter (Metric) Neck:
30 mm
45 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Diameter (Metric) Neck
13681519 2500 mL 45 mm
13681518 1400 mL 30 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13681519 Supplier Fisherbrand™ Supplier No. 2.5 L AMBER RESERVOIR

Side neck allows filling of reservoir without removing the dispenser

  • Amber borosilicate glass
  • Low profile and wide footprint for extra stability

Specifications

Product Type Anti-Tip Reservoir
Autoclavable Not Autoclavable
Color Amber
Material Borosilicate Glass, Nylon Side Filter Neck
Volume (Metric) 2500 mL
For Use With (Equipment) Bottletop Dispenser
Includes Side filler neck, low profile and wide footprint
Volume (English) 84.53 oz.
Diameter (Metric) Neck 45 mm