missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Blank Writable Biohazard Identity Labels

Bright red fluorescent labels display the universal biohazard symbol for easy identification.
$10.83 - $12.14
Specifications
| Certifications/Compliance | OSHA |
|---|---|
| Color | Red |
| Format | Label |
| Material | Paper |
| For Use With (Application) | Biohazard |
| Catalog Number | Mfr. No. | Quantity | Size (English) | Size (Metric) | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Size (English) | Size (Metric) | Price | Quantity & Availability | |||
18999932
![]() |
Fisherbrand™
LBH 20 |
320/Pk. | 0.87 x 3 in. | 2.2 x 7.62 cm |
Each for $10.83
|
|
|||
18999933
![]() |
Fisherbrand™
LBH 21 |
640/Pk. | 0.5 x 1.5 in. | 1.27 x 3.81 cm |
Each for $12.14
|
|
|||
Description
Strong adhesive ensures that labels remain in place.
Compliance
Meet OSHA requirements.
Specifications
| OSHA | |
| Label | |
| Biohazard |
| Red | |
| Paper |