missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Blank Writable Biohazard Identity Labels
GREENER_CHOICE

Catalog No. 18999932
Change view
Click to view available options
Quantity:
320/Pk.
640/Pk.
Size (English):
0.5 x 1.5 in.
0.87 x 3 in.
Size (Metric):
1.27 x 3.81 cm
2.2 x 7.62 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Size (English) Size (Metric)
18999932 320/Pk. 0.87 x 3 in. 2.2 x 7.62 cm
18999933 640/Pk. 0.5 x 1.5 in. 1.27 x 3.81 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18999932 Supplier Fisherbrand™ Supplier No. LBH 20

Bright red fluorescent labels display the universal biohazard symbol for easy identification.

Strong adhesive ensures that labels remain in place.

Compliance

Meet OSHA requirements.

Specifications

Certifications/Compliance OSHA
Color Red
Format Label
Material Paper
Product Type Biohazard Identity Label
For Use With (Application) Biohazard
Quantity 320/Pk.
Size (English) 0.87 x 3 in.
Size (Metric) 2.2 x 7.62 cm