Learn More
Fisherbrand™ Bitran™ Specimen Storage Bags
Choice of Saranex™ odor-control film for long-term storage or polyethylene for short-term storage
Description
Choice of Saranex™ odor-control film for long-term storage or polyethylene for short-term storage
- Use for collecting, transporting and storing tissue specimens
- Minimize storage space and help to eliminate leakage, breakage and contamination
- Resealable double-lock zipper closure
- Designed for storage of tissues and fixatives and for use as mini-anaerobic environments
Chemical Identifiers
| Zip Seal |
Specifications
| Zip Seal | |
| 7 x 8 in. (17.8 x 20.3cm) | |
| 8 in. | |
| 20.3 cm |
| Saranex Film | |
| 7 in. | |
| 17.8 cm |