Learn More
Fisherbrand™ Bitran™ Specimen Storage Bags
Choice of Saranex™ odor-control film for long-term storage or polyethylene for short-term storage
Supplier:  Fisherbrand™ 4740S
 View more versions of this product
Catalog No. 19240093
Only null left
Description
Description
Choice of Saranex™ odor-control film for long-term storage or polyethylene for short-term storage
  • Use for collecting, transporting and storing tissue specimens
  • Minimize storage space and help to eliminate leakage, breakage and contamination
  • Resealable double-lock zipper closure
  • Designed for storage of tissues and fixatives and for use as mini-anaerobic environments
Specifications
Chemical Identifiers
Zip Seal
Specifications
Zip Seal
250
2 in.
5.1 cm
Saranex Film
2 x 4 in. (5.1 x 10.2cm)
4 in.
10.2 cm
Product Suggestions
Product Suggestions