missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Biohazard Specimen Transport Bags
For light-duty use
Description
For light-duty use
- Clear polyethylene with three-wall construction and large printed biohazard graphic
- Reclosable zipper seal
- Cat. Nos. 01-800-02, 01-800-05 and 01-800-06 feature attached document compartment with flap cover that keeps paperwork in place
- Cat. Nos. 01-800-00, 01-800-01, 01-800-03, 01-800-04, 01-800-07, 01-800-08, and 01-800-09 feature attached document compartment without flap cover
- Cat. No. 01-800-05 features a pouch and a 3 × 5 in. (7.6 x 5.1cm) pad impregnated with superabsorbent polymers; pad will absorb 180cc of water, 60cc of urine or 38cc of blood
- Cat. No. 01-800-06 features a scoreline for safe and easy entry without cutting
Compliance
Meet OSHA and NCCLS guidelines
Chemical Identifiers
| OSHA, NCCLS | |
| Clear | |
| Polyethylene |
| Zip Closure | |
| For light-duty use | |
| Biohazard Specimen Transport Bag |
Specifications
| Not Autoclavable | |
| Zip Closure | |
| For light-duty use | |
| Polyethylene | |
| 100 | |
| 6 in. | |
| Biohazard Specimen Transport Bag | |
| 15.24 cm |
| OSHA, NCCLS | |
| Clear | |
| Document compartment | |
| 1000 | |
| 6 x 6 in. (15.24 x 15.24cm) | |
| 6 in. | |
| 1 lb. | |
| 15.24 cm |