missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Biohazard Specimen Transport Bags
For light-duty use
$89.38 - $500.97
Specifications
| Certifications/Compliance | OSHA, NCCLS |
|---|---|
| Closure Type | Zip Closure |
| Color | Clear |
| For Use With (Application) | For light-duty use |
| Material | Polyethylene |
| Catalog Number | Mfr. No. | Dimensions (L x W) | Includes | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | Includes | Price | Quantity & Availability | ||||
|
0180000
|
N/A | 4 x 6 in. (10.16 x 15.24cm) | Document compartment |
Case of 1000 Each for $89.38
|
|
||||
|
0180001
|
N/A | 6 x 6 in. (15.24 x 15.24cm) | Document compartment |
Case of 1000 Each for $194.90
|
|
||||
|
0180004
|
N/A | 9 x 6 in. (22.86 x 15.24cm) | Requisition Pouch, STAT label |
Case of 1000 Each for $173.66
|
|
||||
|
0180005
|
N/A | 9 x 6 in. (22.86 x 15.24cm) | 3 x 5 in. Absorbent Pad |
Case of 500 Each for $131.83
|
|
||||
|
0180007
|
Fisherbrand™
Q604 |
8 x 8 in. (20.3 x 20.32cm) | Document compartment |
Case of 1000 Each for $139.68
|
|
||||
|
0180008
|
N/A | 10 x 8 in. (25.4 x 20.32cm) | Document compartment |
Case of 1000 Each for $229.24
|
|
||||
|
0180009
|
N/A | 12 x 15 in. (30.48 x 38cm) | Document compartment |
Case of 1000 Each for $500.97
|
|
||||
|
0180003
|
N/A | 9 x 6 in. (22.86 x 15.24cm) | Document compartment |
Case of 1000 Each for $157.83
|
|
||||
Description
For light-duty use
- Clear polyethylene with three-wall construction and large printed biohazard graphic
- Reclosable zipper seal
- Cat. Nos. 01-800-02, 01-800-05 and 01-800-06 feature attached document compartment with flap cover that keeps paperwork in place
- Cat. Nos. 01-800-00, 01-800-01, 01-800-03, 01-800-04, 01-800-07, 01-800-08, and 01-800-09 feature attached document compartment without flap cover
- Cat. No. 01-800-05 features a pouch and a 3 x 5 in. (7.6 x 5.1cm) pad impregnated with superabsorbent polymers; pad will absorb 180cc of water, 60cc of urine or 38cc of blood
- Cat. No. 01-800-06 features a scoreline for safe and easy entry without cutting
Compliance
Meet OSHA and NCCLS guidelines
Specifications
| OSHA, NCCLS | |
| Clear | |
| Polyethylene |
| Zip Closure | |
| For light-duty use | |
| Biohazard Specimen Transport Bag |