Learn More
Fisher Science Education™ Bacteria Basic Slide Set
Description
Includes
Plastic box for 25 microscope slides
Set of six slides includes:
- Staphylococcus aureus, pus organism, smear from culture
- Bacteria from mouth, ideal for demonstrating Gram positive and negative bacteria
- Streptococcus lactis, milk souring organism, smear from culture showing short chains
- Escherichia coli, colon bacteria, smear
- Spirillum volutans, a very large spirillum, smear
- Bacillus subtilis, hay bacillus, smear showing bacilli and spores doubly stained
Specifications
Specifications
| Type | Bacteria Basic Slide Set |