Learn More
Fisherbrand™ Amber Glass Dropping Bottles
Ideal for light-sensitive chemicals
$360.89 - $656.64
Specifications
Color Amber
Material Glass
Sterility Non-sterile
Shape Round
Product Type Dropping Bottle
View More Specs
Includes: Screw cap with rubber bulb and glass pipet
Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
02983A
Accessories View Documents Promotion Details
Fisherbrand™
FS15040G30
30 mL
Case of 72 Each for $656.64
Only null left
02983B
Accessories View Documents Promotion Details
Fisherbrand™
FS15040G60
60 mL
Case of 36 Each for $360.89
Only null left
Description
Description
Ideal for light-sensitive chemicals
  • Amber glass protects light-sensitive chemicals
Specifications
Specifications
Amber
Non-sterile
Dropping Bottle
Screw Cap
Glass
Round
Polypropylene
Yes