missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ 384-Well ShallowWell Polypropylene Microplates

Small volume applications
$2,406.75 - $3,221.31
Specifications
| Certifications/Compliance | ANSI |
|---|---|
| Format | Standard |
| Array | 16 x 24 |
| Material | Polypropylene |
| No. of Wells | 384 |
| Catalog Number | Mfr. No. | Color | Sterility | Volume (Metric) Well | For Use With (Application) | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Sterility | Volume (Metric) Well | For Use With (Application) | Price | Quantity & Availability | ||
|
12566615
|
Fisherbrand™
267457 |
White | Non-sterile | 120 μL | Storage or Colorimetric Assays |
Case of 100 Each for $2,585.16
|
|
||
|
12566616
|
Fisherbrand™
267463 |
White | Sterile | 58 μL | Storage or Colorimetric Assays |
Case of 100 Each for $3,221.31
|
|
||
12566617
![]() |
Fisherbrand™
267464 |
Black | Non-sterile | 120 μL | Fluorescent Assays |
Case of 100 Each for $2,406.75
|
|
||
Description
Feature a shared-wall design which yields increased well volume
- Automation compatible, ANSI standard
- Low-binding and chemically resistant surface
- Natural plates for storage or colorimetric assays
- White plates for luminescent assays, including SPA
- Black plates for fluorescent assays
- Round bottom wells for optimal sample recovery
- Rounded square wells minimize wicking (capillary action)
- Working volume range: 2 to 35μL/well
Order Info
Lids not included
Specifications
| ANSI | |
| 16 x 24 | |
| 384 |
| Standard | |
| Polypropylene | |
| Round, Conical Bottom (V-well) |