missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 384-Well ShallowWell Polypropylene Microplates
GREENER_CHOICE

Catalog No. 12566617
Change view
Click to view available options
Color:
Black
White
Sterility:
Non-sterile
Sterile
Volume (Metric) Well:
58 μL
120 μL
For Use With (Application):
Fluorescent Assays
Storage or Colorimetric Assays
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility Volume (Metric) Well For Use With (Application)
12566617 Black Non-sterile 120 μL Fluorescent Assays
12566615 White Non-sterile 120 μL Storage or Colorimetric Assays
12566616 White Sterile 58 μL Storage or Colorimetric Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12566617 Supplier Fisherbrand™ Supplier No. 267464

Small volume applications

Feature a shared-wall design which yields increased well volume

  • Automation compatible, ANSI standard
  • Low-binding and chemically resistant surface
  • Natural plates for storage or colorimetric assays
  • White plates for luminescent assays, including SPA
  • Black plates for fluorescent assays
  • Round bottom wells for optimal sample recovery
  • Rounded square wells minimize wicking (capillary action)
  • Working volume range: 2 to 35μL/well

Order Info

Lids not included

Specifications

Certifications/Compliance ANSI
Format Standard
Array 16 x 24
Color Black
Material Polypropylene
Volume (Metric) Well 120 μL
No. of Wells 384
Sterility Non-sterile
Well Shape Round, Conical Bottom (V-well)
For Use With (Application) Fluorescent Assays