missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Comb for use with FB-VE10-1 Vertical Electrophoresis System
For use with Fisherbrand™ FB-VE10-1 Vertical Electrophoresis System.
$113.43 - $141.42
Specifications
| For Use With (Equipment) | Owl P8 Series Electrophoresis System |
|---|---|
| Type | Comb |
| Gel Dimensions | 10 x 10 cm |
| Catalog Number | Mfr. No. | Thickness (Metric) Teeth | Teeth Count | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Thickness (Metric) Teeth | Teeth Count | Price | Quantity | ||||
|
FB-VE10-19
|
Fisherbrand™
MP8A |
0.77 cm | 8 |
Each for $131.58
|
|
||||
|
FB-VE10-18
|
Fisherbrand™
MP6D |
0.15 cm | 6 |
Each for $131.58
|
|
||||
|
FB-VE10-17
|
Fisherbrand™
MP6C |
0.08 cm | 6 |
Each for $131.58
|
|
||||
|
FB-VE10-20
|
Fisherbrand™
MP8C |
0.08 cm | 8 |
Each for $131.58
|
|
||||
|
FB-VE10-21
|
Fisherbrand™
MP8D |
0.15 cm | 8 |
Each for $131.58
|
|
||||
|
FB-VE10-23
|
Fisherbrand™
MP10C |
0.08 cm | 10 |
Each for $124.84
|
|
||||
|
FB-VE10-22
|
Fisherbrand™
MP10A |
0.05 cm | 10 |
Each for $124.84
|
|
||||
|
FB-VE10-25
|
Fisherbrand™
MP12A |
0.05 cm | 12 |
Each for $113.43
|
|
||||
|
FB-VE10-26
|
Fisherbrand™
MP12C |
0.08 cm | 12 |
Each for $141.42
|
|
||||
|
FB-VE10-29
|
Fisherbrand™
MP20A |
0.05 cm | 20 |
Each for $131.58
|
|
||||
|
FB-VE10-28
|
Fisherbrand™
MP15A |
0.05 cm | 15 |
Each for $131.58
|
|
||||
|
FB-VE10-27
|
Fisherbrand™
MP12D |
0.15 cm | 12 |
Each for $131.58
|
|
||||
|
FB-VE10-24
|
Fisherbrand™
MP10D |
0.15 cm | 10 |
Each for $131.58
|
|
||||
Specifications
| Owl P8 Series Electrophoresis System | |
| Comb | |
| 10 x 10 cm |