Learn More
Fisherbrand™ Vortex Mixer Accessories: Foam Racks
For use with Fisherbrand vortex mixers.
$130.97
Specifications
| Material | Foam |
|---|---|
| For Use With (Equipment) | Digital and analog mini vortex mixer |
Description
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
- Mixes 96-well plates or deepwell blocks
- Requires Insert Retainer (02-215-391)
Specifications
| Foam | |
| Digital and analog mini vortex mixer |