missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Vortex Mixer Accessories: Foam Insert Set
For use with Fisherbrand vortex mixers.
$76.67 - $87.64
| Catalog Number | Mfr. No. | Tube Size | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Tube Size | Price | Quantity & Availability | |||||
|
02215393
|
Fisherbrand™
80862993 |
9 to 13 mm |
Each for $76.67
|
|
|||||
|
02215394
|
Fisherbrand™
80862994 |
9 to 13 mm, 14 to 19 mm, 20 to 29 mm |
Each for $87.64
|
|
|||||
|
02215395
|
Fisherbrand™
80862995 |
20 to 25 mm |
Each for $81.67
|
|
|||||
Description
For use with Fisherbrand vortex mixers.
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
Foam Insert Variety Pack
- Includes foam inserts for the following: 9 to 13mm tube holder, 14 to 19mm tube holder, 20 to 29mm
Specifications
| Foam Insert Set | |
| Foam |