Learn More
Fisherbrand™ Stainless Steel Shaft for Mixing Impellers
Stainless Steel Shaft for Mixing Impellers is composed of 316L stainless steel and passivated for maximum corrosion protection. Available in multiple sizes.
$86.39 - $265.87
| Catalog Number | Mfr. No. | Diameter (Metric) | Diameter (English) | Length (Metric) | Length (English) | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (Metric) | Diameter (English) | Length (Metric) | Length (English) | Price | Quantity & Availability | ||
14500365
![]() |
Fisherbrand™
A712FSR |
8 mm | 0.3125 in. | 300 mm | 12 in. |
Each for $86.39
|
|
||
14500366
![]() |
Fisherbrand™
A722FSR |
8 mm | 0.3125 in. | 457 mm | 18 in. |
Each for $147.23
|
|
||
14500368
![]() |
Fisherbrand™
A733FSR |
9.5 mm | 0.375 in. | 508 mm | 20 in. |
Each for $166.37
|
|
||
14500371
![]() |
Fisherbrand™
A753FSR |
9.5 mm | 0.375 in. | 762 mm | 30 in. |
Each for $265.87
|
|
||
14500367
![]() |
Fisherbrand™
A723FSR |
9.5 mm | 0.375 in. | 457 mm | 18 in. |
Each for $138.65
|
|
||
14500369
![]() |
Fisherbrand™
A742FSR |
8 mm | 0.3125 in. | 610 mm | 24 in. |
Each for $197.06
|
|
||
14500370
![]() |
Fisherbrand™
A743FSR |
9.5 mm | 0.375 in. | 610 mm | 24 in. |
Each for $208.66
|
|
||
Description
- Composed of stainless steel 316L
- 9.5 mm (0.375 in.) shafts have a turned down 8 mm (0.3125 in.) end
- Passivated for corrosion resistance
Specifications
| Impeller Stirrer Shaft | |
| Stainless Steel (316L) |