Learn More
Fisherbrand™ Specific Gravity Hydrometers
Measure true specific gravity of liquids.
Description
Measure true specific gravity of liquids.
Measure true specific gravity of liquids.
- Chemical-resistant bulbs and stems
- Streamlined to avoid trapping air bubbles
- With directional device that stops the instrument with the scale facing the user for more convenient, more accurate readings
Order Info
A certificate of calibration is available; simply add the letter C to the end of the catalog number when ordering.
Compliance
Meet or exceed ASTM™ E 100 for accuracy, ASTM E 126 for calibration
Chemical Identifiers
| Soda Lime Glass |
Specifications
| Specific Gravity Hydrometer | |
| 3-point Cert., ASTM E100, ASTM E126 | |
| 300mL Borosilicate Glass cylinder | |
| Measure true specific gravity of liquids heavier than Water | |
| 250 mL |
| 0.005 | |
| 290 mm | |
| Soda Lime Glass | |
| 1.000 to 1.600 |