missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ MultiTube Vortexers: Centrifuge Tube Racks
Centrifuge tube racks for Fisherbrand™ MultiTube Vortexers.
$120.00 - $124.74
Specifications
| Product Type | Centrifuge Tube Rack |
|---|---|
| For Use With (Equipment) | Multitube vortexer |
Description
Centrifuge tube racks for Fisherbrand™ MultiTube Vortexers.
For use with Fisher Scientific MultiTube Vortexers
Color-coded foam racks for centrifuge tubes
*Color subject to change
Specifications
| Centrifuge Tube Rack | |
| Multitube vortexer |