Learn More
Fisherbrand™ Moticam™ Digital Cameras
Digital microscope camera with Motic software to create still or moving images on computer
$3,874.09
Specifications
Sensor Types CMOS
Warranty 1 Year
View More Specs
Products 1
Catalog Number Mfr. No. Type Pixels Price Quantity & Availability  
Catalog Number Mfr. No. Type Pixels Price Quantity & Availability  
15000109
View Documents Promotion Details
Fisherbrand™
DMOTICAM 1080
Moticam 1080 Digital Microscope Camera 2.2 x 2.2 μm
Each for $3,874.09
Only null left
Description
Description
  • Camera with Motic Images Plus software enables to view, capture, enhance, label, measure, print and store images with one program
  • Available with detachable USB cable
  • Generates live, digital output
  • Operating system compatibility: Microsoft Windows XP/Vista/7; Apple Mac OS
  • Operating temperature: -30° to +70°C non condensing
Specifications
Specifications
CMOS
1 Year