Learn More
Fisherbrand™ High Purity Hydrogen Gas Generators For GC Fuel Gas Accessory, PM Kit
For use with High Purity Hydrogen Gas Generators for GC Fuel Gas.
Supplier:  Fisherbrand™ 604970532L2352
 View more versions of this product
Catalog No. 14950645
Only null left
Includes:
  • Six Month Kit: DI Cartridge, In-line Water filter, and 3 Environmental Tank Vent Fliters
  • 24 Month Kit: Desiccant Cartridge, DI Cartridge, In-line Water filter, 3 Environmental Tank Vent Fliters, Water Pump and Service Dongle
Description
Description
Six month PM Kit
  • Helps ensure quality of deionized water in Fisher Scientifc™ HP range of H2 generators maximizing life of PEM cell
24 month PM Kit
  • Required at 16,000 hours of operation when Fisher Scientific HP generator will alarm indicating that service is due
Specifications
Chemical Identifiers
Gas chromatography, fuel gas, flame ionization detector (GC-FID) flame photometric detector (GC-FPD), Electrolytic Conductivity Detector (ELCD & Hall ELCD), other combustion detectors
High Purity Hydrogen Gas Generators For GC Fuels
Specifications
24 Month PM kit for Fisher Scientific H2 Generator
Gas chromatography, fuel gas, flame ionization detector (GC-FID) flame photometric detector (GC-FPD), Electrolytic Conductivity Detector (ELCD & Hall ELCD), other combustion detectors
1 desiccant cartridge, 1 DI cartridge, 1 in-line water filter and 3 environmental filters, replacement float, water pump and service reset tool
24 Month PM Kit
High Purity Hydrogen Gas Generators For GC Fuels
Product Suggestions
Product Suggestions