Learn More
Fisherbrand™ High Purity Hydrogen Gas Generators For GC Fuel Gas Accessory, Desiccant Cartridge
For use with High Purity Hydrogen Gas Generators for GC Fuel Gas.
Supplier:  Fisherbrand™ 604970412L2352
Catalog No. 14950643
Only null left
Includes: Replacement Desiccant Cartridge
Description
Description
  • Disposable cartridge
  • Final stage in drying for Fisher Scientific™ HP range of hydrogen gas generators
  • Should be replaced whenever indicator beads become opaque to ensure highest gas purity
  • Replaced as needed
Specifications
Specifications
Desiccant Cartridge Kit
Replacement Desiccant Cartridge for Fisher Scientific Hiph Purity H2 Generator
Molecular Sieve MS 544/Clear Polycarbonate
Desiccant cartrigde, wrench and service instructions
High Purity Hydrogen Gas Generators For GC Fuels
Thread
Clear Polycarbonate
EPDM
Gas chromatography, fuel gas, flame ionization detector (GC-FID) flame photometric detector (GC-FPD), Electrolytic Conductivity Detector (ELCD & Hall ELCD), other combustion detectors
Product Suggestions
Product Suggestions