missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Assorted pH Buffer Pouches, Fisherbrand™
Description
Economical, accurate, convenient
- ±0.01pH accuracy at 25°C (77°F)
- Pack of 20 pouches: five pouches each of pH 4, 7, and 10 buffers and rinse solution
Certifications
All solutions are certified traceable to NIST™ standards.
Specifications
Specifications
| For Use With (Application) | Calibration |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 4; 10; 7 |
| Physical Form | Liquid |
| Quantity | 20 pouches |
| Solution Type | Buffer |