missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 850 Homogenizer Accessory, Generator Probe
GREENER_CHOICE

Stainless-steel generator probe; Fisher Scientific™ 850 Homogenizer Accessory, Generator Probe is compatible with Fisher Scientific 850 Homogenizers.

Supplier:  Fisherbrand™ 15340182

 View more versions of this product

Catalog No. 15340182


Only null left

Alert:

Warning: Stainles Steel bearings cannot be autoclaved.


Includes: Generator probe, instruction manual, cleaning brush and spare bearing

Description

Description

  • Comes with PTFE upper and lower bearings
  • Entire probe can be autoclaved
  • Easily disassembled for cleaning

Saw Tooth Generator Probe:

  • Add extra cutting action and stator design allows fine to medium samples to pass through processing windows
Specifications

Chemical Identifiers

Probe
195 mm
316 Stainless Steel
Fisher Scientific 850 Homogenizer

Specifications

Probe
200 mL to 5 L
195 mm
Flat
Fine
30 mm
316 Stainless Steel
Fisher Scientific 850 Homogenizer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions