missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ 850 Homogenizer Accessory, Generator Probe

Stainless-steel generator probe; Fisher Scientific™ 850 Homogenizer Accessory, Generator Probe is compatible with Fisher Scientific 850 Homogenizers.
Supplier: Fisherbrand™ 15340181
Alert:
Warning: Stainles Steel bearings cannot be autoclaved.
Includes: Generator probe, instruction manual, cleaning brush and spare bearing
Description
- Comes with PTFE upper and lower bearings
- Entire probe can be autoclaved
- Easily disassembled for cleaning
Saw Tooth Generator Probe:
- Add extra cutting action and stator design allows fine to medium samples to pass through processing windows
Chemical Identifiers
| Probe | |
| 195 mm | |
| 316 Stainless Steel | |
| Fisher Scientific 850 Homogenizer |
Specifications
| Probe | |
| 100 mL to 2 L | |
| 195 mm | |
| Sawtooth, wide window |
| Coarse | |
| 20 mm | |
| 316 Stainless Steel | |
| Fisher Scientific 850 Homogenizer |